Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172695.1 | NP_001166166.1 | 5 | 693 | |
NM_001172696.1 | NP_001166167.1 | 7 | 844 | ||
NM_005726.5 | NP_005717.3 | 6 | 781 | ||
GenBank mRNA | AF110399.1 | - | 6 | 727 | |
AK295323.1 | - | 8 | 1013 | ||
AK304621.1 | - | 5 | 677 | ||
AK313453.1 | - | 6 | 765 | ||
BC022862.1 | - | 7 | 817 | 4076880 | |
BC093068.1 | - | 6 | 736 | 30722527 | |
L37936.1 | - | 5 | 632 |