Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006472.5 | NP_006463.3 | 1 | 521 | |
XM_017000085.1 | XP_016855574.1 | 1 | 470 | ||
GenBank mRNA | AK226090.1 | - | 1 | 433 | |
AK301148.1 | - | 1 | 43 | ||
AY594328.2 | - | 1 | 156 | ||
BC024305.1 | - | 1 | 50 | ||
BC093702.1 | - | 1 | 230 | 7939547 | |
BC093704.1 | - | 1 | 230 | 7939549 | |
BX537824.1 | - | 1 | 469 | ||
DB058019.1 | - | 1 | 497 | ||
S73591.1 | - | 1 | 377 |