Gene Transcripts

Gene Symbol
BVES
Entrez Gene ID
11149
Gene Name
blood vessel epicardial substance
Gene Aliases
HBVES, LGMD2X, POP1, POPDC1
Chromosome Location
Chr. 6: 105096822 - 105137174 on Build GRCh38
UniGene ID
Hs.221660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199563.1 NP_001186492.1 8 1279
NM_007073.4 NP_009004.2 8 1149
NM_147147.3 NP_671488.1 8 1219
XM_011535398.2 XP_011533700.1 8 1154
GenBank mRNA AF204172.1 - 8 1173
BC034425.1 - 8 1219 4821891
BC040502.1 - 8 1279 5295907

View Full Product Details