Gene Transcripts
- Gene Symbol
- AP4S1
- Entrez Gene ID
- 11154
- Gene Name
- adaptor related protein complex 4 sigma 1 subunit
- Gene Aliases
- AP47B, CLA20, CLAPS4, CPSQ6, SPG52
- Chromosome Location
- Chr. 14: 31025106 - 31096450 on Build GRCh38
- UniGene ID
- Hs.293411
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128126.2 |
NP_001121598.1
|
3, 4 |
618 |
|
NM_001254726.1 |
NP_001241655.1
|
3, 4 |
618 |
|
NM_001254727.1 |
NP_001241656.1
|
3, 4 |
618 |
|
NM_001254728.1 |
NP_001241657.1
|
3, 4 |
975 |
|
NM_001254729.1 |
NP_001241658.1
|
3, 4 |
493 |
|
NM_007077.4 |
NP_009008.2
|
3, 4 |
618 |
|
XM_005267293.4 |
XP_005267350.1
|
4, 5 |
1061 |
|
XM_011536371.2 |
XP_011534673.1
|
4, 5 |
631 |
|
XM_011536372.2 |
XP_011534674.1
|
3, 4 |
1025 |
|
GenBank mRNA |
AB030654.1 |
- |
3, 4 |
438 |
|
AF155159.1 |
- |
3, 4 |
412 |
|
AK304115.1 |
- |
3, 4 |
439 |
|
AK316517.1 |
- |
3, 4 |
441 |
|
BC001259.1 |
- |
3, 4 |
401 |
3454117 |
BF671561.1 |
- |
3, 4 |
491 |
|
BG702035.1 |
- |
3, 4 |
627 |
|
BT006701.1 |
- |
2, 3 |
229 |
|
BX247969.1 |
- |
3, 4 |
420 |
|
BX388185.2 |
- |
3, 4 |
404 |
|
CR457100.1 |
- |
2, 3 |
229 |
|
View Full Product Details