Gene Transcripts

Gene Symbol
OR56A1
Entrez Gene ID
120796
Gene Name
olfactory receptor family 56 subfamily A member 1
Gene Aliases
OR11-75
Chromosome Location
Chr. 11: 6026671 - 6027741 on Build GRCh38
UniGene ID
Hs.553543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001917.2 NP_001001917.2 1 892
GenBank mRNA BC136904.1 - 1 892
BC136911.1 - 1 892 9020902

View Full Product Details