Gene Transcripts

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
Chromosome Location
Chr. 14: 51489100 - 51730727 on Build GRCh38
UniGene ID
Hs.434914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042481.2 NP_001035946.1 4 680
NM_001267046.1 NP_001253975.1 3 456
NM_152330.3 NP_689543.1 3 456
XM_006720029.1 XP_006720092.1 3 553
XM_006720030.1 XP_006720093.1 3 553
XM_011536423.1 XP_011534725.1 4 700
XM_011536424.1 XP_011534726.1 5 825
XM_017020967.1 XP_016876456.1 3 406
XM_017020968.1 XP_016876457.1 3 406
GenBank mRNA AK055545.1 - 3 367
AK126521.1 - 1 101
AK315764.1 - 3 377
AL833158.1 - 4 666
BC020521.2 - 3 404 3914276
BX161430.1 - 3 456
BX647863.1 - 3 368

View Full Product Details