Gene Transcripts
- Gene Symbol
- SLC9B1
- Entrez Gene ID
- 150159
- Gene Name
- solute carrier family 9 member B1
- Gene Aliases
- NHA1, NHEDC1
- Chromosome Location
- Chr. 4: 102885048 - 103019739 on Build GRCh38
- UniGene ID
- Hs.666728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100874.2 |
NP_001094344.1
|
2, 3 |
219 |
|
NM_139173.3 |
NP_631912.2
|
2, 3 |
219 |
|
NR_047513.1 |
|
2, 3 |
219 |
|
NR_047515.1 |
|
2, 3 |
219 |
|
XM_006714093.3 |
XP_006714156.1
|
3, 4 |
238 |
|
XM_011531622.1 |
XP_011529924.1
|
3, 4 |
202 |
|
XM_011531623.1 |
XP_011529925.1
|
2, 3 |
206 |
|
GenBank mRNA |
AF447585.1 |
- |
2, 3 |
199 |
|
AK313592.1 |
- |
2, 3 |
146 |
|
AY461581.1 |
- |
2, 3 |
167 |
|
BC022079.1 |
- |
2, 3 |
135 |
|
BC046636.1 |
- |
2, 3 |
143 |
|
BC110794.1 |
- |
2, 3 |
103 |
5745227 |
BC132712.1 |
- |
2, 3 |
146 |
40146734 |
BC136966.1 |
- |
2, 3 |
146 |
9020958 |
DQ003059.1 |
- |
2, 3 |
199 |
|
View Full Product Details