Gene Transcripts

Gene Symbol
OR1Q1
Entrez Gene ID
158131
Gene Name
olfactory receptor family 1 subfamily Q member 1
Gene Aliases
HSTPCR106, OR1Q2, OR1Q3, OR9-25, OR9-A, OST226, OST226OR9-A, TPCR106
Chromosome Location
Chr. 9: 122614738 - 122615682 on Build GRCh38
UniGene ID
Hs.684864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012364.1 NP_036496.1 1 292
GenBank mRNA BC140713.1 - 1 361

View Full Product Details