Gene Transcripts

Gene Symbol
OR5B21
Entrez Gene ID
219968
Gene Name
olfactory receptor family 5 subfamily B member 21
Gene Aliases
Chromosome Location
Chr. 11: 58507176 - 58508105 on Build GRCh38
UniGene ID
Hs.553647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005218.1 NP_001005218.1 1 134

View Full Product Details