Gene Transcripts
- Gene Symbol
- RPH3A
- Entrez Gene ID
- 22895
- Gene Name
- rabphilin 3A
- Gene Aliases
- Chromosome Location
- Chr. 12: 112791538 - 112898881 on Build GRCh38
- UniGene ID
- Hs.21239
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK295696.1 |
- |
17 |
1830 |
|
BC017259.2 |
- |
16 |
1671 |
3510158 |
View Full Product Details