Gene Transcripts

Gene Symbol
PDS5A
Entrez Gene ID
23244
Gene Name
PDS5 cohesin associated factor A
Gene Aliases
PIG54, SCC-112, SCC112
Chromosome Location
Chr. 4: 39822863 - 39977956 on Build GRCh38
UniGene ID
Hs.331431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100399.1 NP_001093869.1 14, 15 2123
NM_001100400.1 NP_001093870.1 14, 15 2123
XM_011513672.2 XP_011511974.1 14, 15 1736
XM_017007928.1 XP_016863417.1 14, 15 1698
GenBank mRNA AB014548.1 - 2, 3 127
AB621803.1 - 14, 15 2137
AF294791.1 - 14, 15 1694
AK098331.1 - 14, 15 1707
AY550968.1 - 13, 14 1620
BC041361.1 - 14, 15 2127
BC114218.1 - 14, 15 1819 5579832
BC126225.1 - 13, 14 1533 8991941
CA412233.1 - 2, 3 619
CR749425.1 - 13, 14 1492

View Full Product Details