Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005267.4 | NP_005258.2 | 2 | 525 | |
XM_011509416.1 | XP_011507718.1 | 1 | 1272 | ||
XM_011509417.1 | XP_011507719.1 | 1 | 1272 | ||
GenBank mRNA | DB476698.1 | - | 1 | 162 | |
KJ534845.1 | - | 1 | 472 |