Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014026.4 | NP_054745.1 | 6 | 1151 | |
XM_011542778.2 | XP_011541080.1 | 6 | 880 | ||
XM_011542780.2 | XP_011541082.1 | 6 | 948 | ||
XM_017017612.1 | XP_016873101.1 | 7 | 1845 | ||
GenBank mRNA | AF077201.1 | - | 6 | 834 | |
AF532613.1 | - | 6 | 1151 | ||
AK026554.1 | - | 6 | 847 | ||
AK090828.1 | - | 3 | 2172 | ||
AK223089.1 | - | 6 | 847 | ||
AK307403.1 | - | 6 | 843 | ||
AY040771.1 | - | 6 | 849 | ||
AY077684.1 | - | 6 | 822 | ||
BC014532.1 | - | 6 | 844 | 4064215 |