Gene Transcripts

Gene Symbol
UBIAD1
Entrez Gene ID
29914
Gene Name
UbiA prenyltransferase domain containing 1
Gene Aliases
SCCD, TERE1
Chromosome Location
Chr. 1: 11273198 - 11299578 on Build GRCh38
UniGene ID
Hs.522933
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013319.2 NP_037451.1 1 633
XM_006710590.3 XP_006710653.1 1 633
XM_011541304.2 XP_011539606.1 1 633
GenBank mRNA AF117064.1 - 1 631
AK074890.1 - 1 625
AK222808.1 - 1 633
BC004468.2 - 1 325 3936638
BT006832.1 - 1 299

View Full Product Details