Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001015508.2 | NP_001015508.1 | 1 | 1276 | |
NM_001323311.1 | NP_001310240.1 | 2 | 466 | ||
NM_001323312.1 | NP_001310241.1 | 2 | 466 | ||
NM_013357.2 | NP_037489.1 | 1 | 362 | ||
GenBank mRNA | AF195513.2 | - | 1 | 1276 | |
AY077841.1 | - | 1 | 1276 | ||
BC106708.2 | - | 1 | 362 | 40005076 | |
DR003356.1 | - | 2 | 471 |