Gene Transcripts

Gene Symbol
TAAR6
Entrez Gene ID
319100
Gene Name
trace amine associated receptor 6
Gene Aliases
TA4, TAR4, TAR6, TRAR4, taR-4, taR-6
Chromosome Location
Chr. 6: 132570322 - 132571359 on Build GRCh38
UniGene ID
Hs.434196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175067.1 NP_778237.1 1 802
GenBank mRNA AY183470.1 - 1 802
BC069157.1 - 1 802 7217018

View Full Product Details