Gene Transcripts

Gene Symbol
ZDHHC21
Entrez Gene ID
340481
Gene Name
zinc finger DHHC-type containing 21
Gene Aliases
9130404H11Rik, DHHC-21, DHHC21, DNZ1, HSPC097
Chromosome Location
Chr. 9: 14588797 - 14693531 on Build GRCh38
UniGene ID
Hs.649522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178566.4 NP_848661.1 10 1261
XM_017014682.1 XP_016870171.1 8 1045
XM_017014683.1 XP_016870172.1 11 1301
XM_017014684.1 XP_016870173.1 11 1233
XM_017014685.1 XP_016870174.1 11 1181
XM_017014686.1 XP_016870175.1 12 1362
XM_017014687.1 XP_016870176.1 11 1309
XM_017014688.1 XP_016870177.1 11 1391
XM_017014689.1 XP_016870178.1 11 1367
XM_017014690.1 XP_016870179.1 12 1251
XM_017014691.1 XP_016870180.1 11 3776
XM_017014692.1 XP_016870181.1 11 1041
XM_017014693.1 XP_016870182.1 12 1098
GenBank mRNA AK096860.1 - 3 266
AK127313.1 - 3 1524
AK292960.1 - 10 1068
BC042558.1 - 10 1255 4837003

View Full Product Details