Gene Transcripts

Gene Symbol
DPPA3
Entrez Gene ID
359787
Gene Name
developmental pluripotency associated 3
Gene Aliases
STELLA
Chromosome Location
Chr. 12: 7711454 - 7717559 on Build GRCh38
UniGene ID
Hs.131358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199286.3 NP_954980.1 2 301
GenBank mRNA AY317075.1 - 2 301

View Full Product Details