Gene Transcripts
- Gene Symbol
- EIF3E
- Entrez Gene ID
- 3646
- Gene Name
- eukaryotic translation initiation factor 3 subunit E
- Gene Aliases
- EIF3-P48, EIF3S6, INT6, eIF3-p46
- Chromosome Location
- Chr. 8: 108201743 - 108248802 on Build GRCh38
- UniGene ID
- Hs.405590
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001568.2 |
NP_001559.1
|
10, 11 |
1091 |
|
XM_017013398.1 |
XP_016868887.1
|
10, 11 |
1163 |
|
GenBank mRNA |
AK124178.1 |
- |
10, 11 |
1695 |
|
AK313186.1 |
- |
10, 11 |
1080 |
|
BC000734.2 |
- |
10, 11 |
1067 |
2959855 |
BC005944.1 |
- |
10, 11 |
1080 |
|
BC008419.1 |
- |
10, 11 |
1085 |
4080571 |
BC016706.1 |
- |
10, 11 |
1082 |
4096434 |
BC017887.1 |
- |
10, 11 |
1087 |
4685684 |
BC021679.1 |
- |
10, 11 |
1082 |
4132737 |
CR457029.1 |
- |
10, 11 |
1063 |
|
CR542275.1 |
- |
10, 11 |
1063 |
|
U54562.1 |
- |
10, 11 |
1069 |
|
U62962.1 |
- |
10, 11 |
1085 |
|
U85947.1 |
- |
10, 11 |
1071 |
|
U94175.1 |
- |
10, 11 |
1083 |
|
View Full Product Details