Gene Transcripts
- Gene Symbol
- MUC21
- Entrez Gene ID
- 394263
- Gene Name
- mucin 21, cell surface associated
- Gene Aliases
- C6orf205, KMQK697, MUC-21
- Chromosome Location
- Chr. 6: 30983708 - 30988637 on Build GRCh38
- UniGene ID
- Hs.448277
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001010909.3 |
NP_001010909.2
|
1, 2 |
316 |
|
NM_001322370.1 |
NP_001309299.1
|
|
292 |
|
NM_001322371.1 |
NP_001309300.1
|
|
309 |
|
GenBank mRNA |
AB242595.1 |
- |
1, 2 |
226 |
|
AK298642.1 |
- |
1, 2 |
304 |
|
AK299471.1 |
- |
|
309 |
|
AK316363.1 |
- |
1, 2 |
306 |
|
AY358415.1 |
- |
|
303 |
|
BC105735.1 |
- |
1, 2 |
251 |
40026480 |
BC105737.1 |
- |
1, 2 |
251 |
40026476 |
BC107478.1 |
- |
1, 2 |
251 |
40026469 |
DA763771.1 |
- |
1, 2 |
316 |
|
DC373882.1 |
- |
1, 2 |
306 |
|
HM596784.1 |
- |
1, 2 |
245 |
|
View Full Product Details