Gene Transcripts

Gene Symbol
ANKHD1-EIF4EBP3
Entrez Gene ID
404734
Gene Name
ANKHD1-EIF4EBP3 readthrough
Gene Aliases
MASK-BP3, MASK-BP3ARF
Chromosome Location
Chr. 5: 140401814 - 140549578 on Build GRCh38
UniGene ID
Hs.594084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020690.5 NP_065741.3 34 7785
GenBank mRNA AB029008.1 - 5 4891
AF521883.1 - 34 7691
AK125734.1 - 7 1749
BC144623.1 - 34 7870

View Full Product Details