Gene Transcripts

Gene Symbol
PBX1
Entrez Gene ID
5087
Gene Name
PBX homeobox 1
Gene Aliases
Chromosome Location
Chr. 1: 164559360 - 164886047 on Build GRCh38
UniGene ID
Hs.557097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204963.1 NP_001191892.1 6, 7 1460
NM_002585.3 NP_002576.1 6, 7 1460
XM_005245228.3 XP_005245285.1 7, 8 1628
XM_005245229.3 XP_005245286.1 7, 8 1221
XM_011509590.1 XP_011507892.1 7, 8 1628
XM_017001395.1 XP_016856884.1 6, 7 1460
GenBank mRNA AK299673.1 - 6, 7 1076
AK301540.1 - 5, 6 805
BC101578.1 - 6, 7 1049 8069084
BC143435.1 - 5, 6 975
M31170.1 - 4, 5 733
M86546.1 - 6, 7 1120

View Full Product Details