Gene Transcripts
- Gene Symbol
- UBXN1
- Entrez Gene ID
- 51035
- Gene Name
- UBX domain protein 1
- Gene Aliases
- 2B28, SAKS1, UBXD10
- Chromosome Location
- Chr. 11: 62676498 - 62679117 on Build GRCh38
- UniGene ID
- Hs.351296
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286077.1 |
NP_001273006.1
|
5, 6 |
667 |
|
NM_001286078.1 |
NP_001273007.1
|
3, 4 |
490 |
|
NM_015853.4 |
NP_056937.2
|
5, 6 |
667 |
|
XM_005274033.4 |
XP_005274090.1
|
6, 7 |
580 |
|
XM_011545090.2 |
XP_011543392.1
|
5, 6 |
626 |
|
XM_017017874.1 |
XP_016873363.1
|
6, 7 |
576 |
|
GenBank mRNA |
AK054832.1 |
- |
4, 5 |
744 |
|
AK297856.1 |
- |
5, 6 |
645 |
|
AK303471.1 |
- |
4, 5 |
526 |
|
AK316012.1 |
- |
5, 6 |
617 |
|
BC000902.2 |
- |
5, 6 |
606 |
2900493 |
BC001372.2 |
- |
5, 6 |
605 |
3051213 |
BC032689.1 |
- |
4, 5 |
729 |
|
BC040129.1 |
- |
5, 6 |
642 |
6065505 |
BI855821.1 |
- |
3, 4 |
433 |
|
M68864.1 |
- |
5, 6 |
611 |
|
View Full Product Details