Gene Transcripts

Gene Symbol
TRIM17
Entrez Gene ID
51127
Gene Name
tripartite motif containing 17
Gene Aliases
RBCC, RNF16, terf
Chromosome Location
Chr. 1: 228407478 - 228416882 on Build GRCh38
UniGene ID
Hs.121748
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024940.2 NP_001020111.1 5, 6 1135
NM_001134855.1 NP_001128327.1 5, 6 1287
NM_016102.3 NP_057186.1 5, 6 1171
XM_006711779.2 XP_006711842.1 5, 6 1198
XM_011544209.2 XP_011542511.1 5, 6 1185
XM_011544210.2 XP_011542512.1 5, 6 1287
XM_011544211.2 XP_011542513.1 5, 6 1188
GenBank mRNA AF156271.1 - 5, 6 894
AK301105.1 - 5, 6 1091
AK308774.1 - 5, 6 1162
BC033788.1 - 5, 6 935 5165609

View Full Product Details