Gene Transcripts
- Gene Symbol
- LARP7
- Entrez Gene ID
- 51574
- Gene Name
- La ribonucleoprotein domain family member 7
- Gene Aliases
- ALAZS, HDCMA18P, PIP7S
- Chromosome Location
- Chr. 4: 112636964 - 112657592 on Build GRCh38
- UniGene ID
- Hs.713663
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267039.1 |
NP_001253968.1
|
5, 6 |
797 |
|
NM_015454.2 |
NP_056269.1
|
3, 4 |
461 |
|
NM_016648.3 |
NP_057732.2
|
3, 4 |
589 |
|
NR_049768.1 |
|
3, 4 |
486 |
|
GenBank mRNA |
AF068284.1 |
- |
1, 2 |
36 |
|
AK000274.1 |
- |
3, 4 |
399 |
|
BC006981.1 |
- |
3, 4 |
373 |
|
BC066945.1 |
- |
5, 6 |
635 |
4837780 |
BX647100.1 |
- |
3, 4 |
323 |
|
CN346208.1 |
- |
3, 4 |
468 |
|
EU667388.1 |
- |
3, 4 |
332 |
|
View Full Product Details