Gene Transcripts
- Gene Symbol
- ERGIC3
- Entrez Gene ID
- 51614
- Gene Name
- ERGIC and golgi 3
- Gene Aliases
- C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
- Chromosome Location
- Chr. 20: 35542029 - 35557634 on Build GRCh38
- UniGene ID
- Hs.472558
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015966.2 |
NP_057050.1
|
3, 4 |
308 |
|
NM_198398.1 |
NP_938408.1
|
3, 4 |
308 |
|
GenBank mRNA |
AF077030.1 |
- |
3, 4 |
266 |
|
AF116614.1 |
- |
3, 4 |
415 |
|
AF151812.1 |
- |
3, 4 |
266 |
|
AF308298.1 |
- |
3, 4 |
268 |
|
AK074550.1 |
- |
3, 4 |
256 |
|
AK096484.1 |
- |
3, 4 |
256 |
|
AK300916.1 |
- |
3, 4 |
257 |
|
BC009765.1 |
- |
3, 4 |
262 |
4283844 |
BC014014.2 |
- |
3, 4 |
261 |
3954269 |
BU528333.1 |
- |
3, 4 |
285 |
|
CA489637.1 |
- |
3, 4 |
308 |
|
EF206690.1 |
- |
3, 4 |
266 |
|
View Full Product Details