Gene Transcripts

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
Chromosome Location
Chr. 20: 35542029 - 35557634 on Build GRCh38
UniGene ID
Hs.472558
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015966.2 NP_057050.1 6 592
NM_198398.1 NP_938408.1 6 592
GenBank mRNA AF077030.1 - 6 550
AF091085.1 - 4 203
AF116614.1 - 6 699
AF151812.1 - 6 550
AF308298.1 - 6 552
AK074550.1 - 6 540
AK300916.1 - 6 541
AL161963.1 - 3 233
BC009765.1 - 6 546 4283844
BC014014.2 - 6 545 3954269
BU528333.1 - 6 569
CA489637.1 - 6 592
EF206690.1 - 6 550

View Full Product Details