Gene Transcripts

Gene Symbol
PEX7
Entrez Gene ID
5191
Gene Name
peroxisomal biogenesis factor 7
Gene Aliases
PBD9B, PTS2R, RCDP1, RD
Chromosome Location
Chr. 6: 136822220 - 136913934 on Build GRCh38
UniGene ID
Hs.280932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000288.3 NP_000279.1 9 971
XM_005267019.4 XP_005267076.1 9 1251
XM_006715502.1 XP_006715565.1 6 1021
XM_017010934.1 XP_016866423.1 6 1038
GenBank mRNA BC006268.1 - 9 913
BC031606.1 - 10 1063 5176358
CR542054.1 - 9 869
CR542076.1 - 9 869
U76560.1 - 9 917
U88871.1 - 9 950

View Full Product Details