Gene Transcripts

Gene Symbol
UGT1A4
Entrez Gene ID
54657
Gene Name
UDP glucuronosyltransferase family 1 member A4
Gene Aliases
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D
Chromosome Location
Chr. 2: 233718792 - 233773299 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007120.2 NP_009051.1 1 390
GenBank mRNA AK313623.1 - 1 404
AY435139.1 - 1 361
BC121036.1 - 1 361 40117585
BC131623.1 - 1 354
BC139784.1 - 1 381 40117089
DQ364249.1 - 1 370
M57951.1 - 1 390

View Full Product Details