Gene Transcripts

Gene Symbol
PPP2R5A
Entrez Gene ID
5525
Gene Name
protein phosphatase 2 regulatory subunit B'alpha
Gene Aliases
B56A, PR61A
Chromosome Location
Chr. 1: 212285537 - 212361863 on Build GRCh38
UniGene ID
Hs.744012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199756.1 NP_001186685.1 11 1293
NM_006243.3 NP_006234.1 11 1761
GenBank mRNA AK302202.1 - 11 1293
AK312530.1 - 12 1576
BC022474.1 - 11 1347 4815091
BC110883.1 - 11 1470 6045358
L42373.1 - 11 1758

View Full Product Details