Gene Transcripts

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
Chromosome Location
Chr. 10: 50184861 - 50251564 on Build GRCh38
UniGene ID
Hs.512645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143974.1 NP_001137446.1 2, 3 354
NM_019893.2 NP_063946.2 2, 3 354
XM_011539970.2 XP_011538272.1 3, 4 550
XM_011539971.2 XP_011538273.1 3, 4 554
XM_011539972.2 XP_011538274.1 3, 4 551
XM_011539973.2 XP_011538275.1 3, 4 560
XM_017016435.1 XP_016871924.1 3, 4 554
GenBank mRNA AF250847.1 - 2, 3 297
AF449759.2 - 2, 3 354
BC107105.2 - 2, 3 297 40018929

View Full Product Details