Gene Transcripts

Gene Symbol
PSG6
Entrez Gene ID
5675
Gene Name
pregnancy specific beta-1-glycoprotein 6
Gene Aliases
PSBG-10, PSBG-12, PSBG-6, PSG10, PSGGB
Chromosome Location
Chr. 19: 42902082 - 42917924 on Build GRCh38
UniGene ID
Hs.646353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031850.3 NP_001027020.1 2 235
NM_002782.4 NP_002773.1 2 235
GenBank mRNA BC020652.1 - 2 202
DA833634.1 - 2 235
M31125.1 - 2 148
M33666.1 - 2 155
X17098.1 - 2 156

View Full Product Details