Gene Transcripts

Gene Symbol
RGMA
Entrez Gene ID
56963
Gene Name
repulsive guidance molecule family member a
Gene Aliases
RGM
Chromosome Location
Chr. 15: 93043406 - 93089214 on Build GRCh38
UniGene ID
Hs.271277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166283.1 NP_001159755.1 3 570
NM_001166286.1 NP_001159758.1 2 430
NM_001166287.1 NP_001159759.1 3 544
NM_001166288.1 NP_001159760.1 3 428
NM_001166289.1 NP_001159761.1 3 448
NM_020211.2 NP_064596.2 3 597
GenBank mRNA AK074910.1 - 3 587
AK074966.1 - 2 404
AK074980.1 - 3 364
AK125204.1 - 4 553
AK291365.1 - 2 227
AK309814.1 - 4 810
AL136826.1 - 3 411
AL390083.1 - 2 353
BC015886.1 - 2 421
BC140838.1 - 3 375 9021706
BC151132.1 - 3 375 9057032
DA295545.1 - 3 446
DA512691.1 - 3 428

View Full Product Details