Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000986.3 | NP_000977.1 | 3 | 195 | |
GenBank mRNA | AK311992.1 | - | 3 | 199 | |
BC000690.2 | - | 3 | 174 | 3349215 | |
BC070193.1 | - | 3 | 172 | 6733162 | |
BU754101.1 | - | 3 | 203 | ||
CD513036.1 | - | 2 | 181 | ||
CR456729.1 | - | 2 | 152 | ||
EU668341.1 | - | 3 | 174 | ||
M94314.1 | - | 3 | 191 |