Gene Transcripts

Gene Symbol
TMEM72
Entrez Gene ID
643236
Gene Name
transmembrane protein 72
Gene Aliases
C10orf127, KSP37
Chromosome Location
Chr. 10: 44911307 - 44937010 on Build GRCh38
UniGene ID
Hs.292788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123376.2 NP_001116848.1 1 217
XM_011540071.2 XP_011538373.1 1 226
GenBank mRNA AB235418.1 - 1 172
BC127789.1 - 1 217 40133552
BC127790.1 - 1 217 40133554
BX538120.1 - 352

View Full Product Details