Gene Transcripts

Gene Symbol
SLC1A4
Entrez Gene ID
6509
Gene Name
solute carrier family 1 member 4
Gene Aliases
ASCT1, SATT, SPATCCM
Chromosome Location
Chr. 2: 64988445 - 65023865 on Build GRCh38
UniGene ID
Hs.654352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003038.4 NP_003029.2 5 1189
XM_006712079.2 XP_006712142.1 5 555
GenBank mRNA AB026689.1 - 5 927
AK126636.1 - 4 2379
AK223486.1 - 5 1151
AK298472.1 - 6 1100
AK313052.1 - 5 1153
AK315986.1 - 6 1111
BC026216.1 - 5 1090 4420405
BC072423.1 - 5 1149 6186177
DA218921.1 - 5 557
L14595.1 - 5 1089
L19444.1 - 5 1068

View Full Product Details