Gene Transcripts

Gene Symbol
TCN1
Entrez Gene ID
6947
Gene Name
transcobalamin 1
Gene Aliases
HC, TC-1, TC1, TCI
Chromosome Location
Chr. 11: 59852808 - 59866568 on Build GRCh38
UniGene ID
Hs.2012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001062.3 NP_001053.2 3 378
GenBank mRNA AK292990.1 - 3 295
BC018632.1 - 3 300
BP370727.1 - 3 384
J05068.1 - 3 355

View Full Product Details