Gene Transcripts

Gene Symbol
TM4SF4
Entrez Gene ID
7104
Gene Name
transmembrane 4 L six family member 4
Gene Aliases
ILTMP, il-TMP
Chromosome Location
Chr. 3: 149474581 - 149503394 on Build GRCh38
UniGene ID
Hs.133527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004617.3 NP_004608.1 1 368
GenBank mRNA AK055577.1 - 1 975
AK315464.1 - 1 255
BC001386.2 - 1 238 3051146
BP294159.1 - 1 368
BT007430.1 - 1 71
U31449.1 - 1 236

View Full Product Details