Gene Transcripts

Gene Symbol
CEACAM18
Entrez Gene ID
729767
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
Gene Aliases
Chromosome Location
Chr. 19: 51478543 - 51491261 on Build GRCh38
UniGene ID
Hs.653111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278392.1 NP_001265321.1 2, 3 390
XM_011527258.2 XP_011525560.1 2, 3 490

View Full Product Details