Gene Transcripts

Gene Symbol
C1orf54
Entrez Gene ID
79630
Gene Name
chromosome 1 open reading frame 54
Gene Aliases
Chromosome Location
Chr. 1: 150272284 - 150280927 on Build GRCh38
UniGene ID
Hs.91283
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301039.1 NP_001287968.1 2 176
NM_001301040.1 NP_001287969.1 2 176
NM_001301041.1 NP_001287970.1 1 46
NM_001301042.1 NP_001287971.1 1 46
NM_024579.3 NP_078855.2 1 65
XM_011509984.2 XP_011508286.1 2 176
GenBank mRNA AA095731.1 - 1 33
AI342055.1 - 5 350
AK026874.1 - 1 50
AX770997.1 - 1 63
AX770998.1 - 2 90
AX770999.1 - 1 65
BC017761.1 - 1 50 4617936
DA358536.1 - 1 57
T82829.1 - 2 156

View Full Product Details