Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024761.4 | NP_079037.3 | 2 | 511 | |
GenBank mRNA | AJ580636.1 | - | 1 | 15 | |
AK023266.1 | - | 2 | 440 | ||
AL832572.2 | - | 2 | 405 | ||
AU126938.1 | - | 2 | 403 | ||
BC033027.1 | - | 2 | 423 | 5268356 | |
CR457307.1 | - | 1 | 15 | ||
DB266074.1 | - | 2 | 440 |