Gene Transcripts

Gene Symbol
CALU
Entrez Gene ID
813
Gene Name
calumenin
Gene Aliases
Chromosome Location
Chr. 7: 128739292 - 128773423 on Build GRCh38
UniGene ID
Hs.743262
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199671.1 NP_001186600.1 4 1326
NM_001199673.1 NP_001186602.1 3 511
NM_001219.4 NP_001210.1 3 511
XM_017012659.1 XP_016868148.1 3 876
GenBank mRNA AF013759.1 - 3 417
AK056338.1 - 4 1214
AK074862.1 - 3 461
AK312446.1 - 3 454
BC013383.2 - 3 410 3507961
BT006826.1 - 2 359
BT020143.1 - 2 359
CR457039.1 - 2 359
CR541835.1 - 2 359
DA023987.1 - 3 511
HM002604.1 - 4 1327
HM002606.1 - 3 511
HM002615.1 - 4 1327
U67280.1 - 3 421

View Full Product Details