Gene Transcripts
- Gene Symbol
- CDT1
- Entrez Gene ID
- 81620
- Gene Name
- chromatin licensing and DNA replication factor 1
- Gene Aliases
- DUP, RIS2
- Chromosome Location
- Chr. 16: 88803778 - 88809258 on Build GRCh38
- UniGene ID
- Hs.122908
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030928.3 |
NP_112190.2
|
7, 8 |
1183 |
|
GenBank mRNA |
AB053172.1 |
- |
7, 8 |
1183 |
|
AF070552.1 |
- |
5, 6 |
722 |
|
AF321125.1 |
- |
7, 8 |
1154 |
|
BC000137.2 |
- |
7, 8 |
1145 |
3048353 |
BC008676.1 |
- |
7, 8 |
1021 |
|
BC008860.2 |
- |
7, 8 |
1076 |
|
BC009410.1 |
- |
7, 8 |
1139 |
3613566 |
BC014202.2 |
- |
3, 4 |
444 |
|
BC021126.1 |
- |
5, 6 |
715 |
|
BC049205.1 |
- |
5, 6 |
787 |
|
View Full Product Details