Gene Transcripts

Gene Symbol
TRIM7
Entrez Gene ID
81786
Gene Name
tripartite motif containing 7
Gene Aliases
GNIP, RNF90
Chromosome Location
Chr. 5: 181193924 - 181206000 on Build GRCh38
UniGene ID
Hs.487412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203293.2 NP_976038.1 5 1098
NM_203294.1 NP_976039.1 5 712
NM_203295.1 NP_976040.1 5 780
NM_203296.1 NP_976041.1 5 989
NM_203297.1 NP_976042.1 3 1146
XM_017009903.1 XP_016865392.1 5 1137
GenBank mRNA AF396651.1 - 5 975
AF396652.1 - 5 712
AF396653.1 - 5 780
AF396654.1 - 5 773
AF396655.1 - 3 1152
BC132863.1 - 5 977 40146885
BC132867.1 - 5 977 40146889

View Full Product Details