Gene Transcripts
- Gene Symbol
- CHST9
- Entrez Gene ID
- 83539
- Gene Name
- carbohydrate sulfotransferase 9
- Gene Aliases
- GALNAC4ST-2, GalNAc4ST2
- Chromosome Location
- Chr. 18: 26908754 - 27185364 on Build GRCh38
- UniGene ID
- Hs.44584
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256316.1 |
NP_001243245.1
|
2, 3 |
424 |
|
NM_031422.5 |
NP_113610.2
|
2, 3 |
424 |
|
XM_006722555.3 |
XP_006722618.1
|
2, 3 |
381 |
|
XM_017026033.1 |
XP_016881522.1
|
1, 2 |
323 |
|
XM_017026034.1 |
XP_016881523.1
|
2, 3 |
450 |
|
GenBank mRNA |
AF239821.1 |
- |
2, 3 |
301 |
|
AF332472.1 |
- |
1, 2 |
215 |
|
AJ707539.1 |
- |
2, 3 |
411 |
|
AK093349.1 |
- |
1, 2 |
323 |
|
AY358488.1 |
- |
2, 3 |
403 |
|
BC025764.1 |
- |
2, 3 |
368 |
5205829 |
BM919514.1 |
- |
2, 3 |
403 |
|
DB444451.1 |
- |
1, 2 |
354 |
|
KU178792.1 |
- |
1, 2 |
110 |
|
KU178793.1 |
- |
1, 2 |
110 |
|
View Full Product Details