Gene Transcripts

Gene Symbol
HIST1H4L
Entrez Gene ID
8368
Gene Name
histone cluster 1, H4l
Gene Aliases
H4.k, H4/k, H4FK
Chromosome Location
Chr. 6: 27873148 - 27873511 on Build GRCh38
UniGene ID
Hs.533295
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003546.2 NP_003537.1 1 172
GenBank mRNA AB451459.1 - 1 171
BC069392.1 - 1 172 7262681

View Full Product Details