Gene Transcripts

Gene Symbol
NT5C1A
Entrez Gene ID
84618
Gene Name
5'-nucleotidase, cytosolic IA
Gene Aliases
CN-I, CN-IA, CN1, CN1A, CNI
Chromosome Location
Chr. 1: 39659121 - 39672038 on Build GRCh38
UniGene ID
Hs.307006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032526.2 NP_115915.1 3 401
GenBank mRNA AF331801.1 - 3 401
AY028778.1 - 3 392
BC103879.1 - 3 401 40004489
BC103880.1 - 3 401 40004491

View Full Product Details