Gene Transcripts

Gene Symbol
UBE2Q2
Entrez Gene ID
92912
Gene Name
ubiquitin conjugating enzyme E2 Q2
Gene Aliases
Chromosome Location
Chr. 15: 75843286 - 75901047 on Build GRCh38
UniGene ID
Hs.23033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145335.1 NP_001138807.1 3 558
NM_173469.3 NP_775740.1 3 677
XM_005254787.1 XP_005254844.1 3 449
XM_005254788.1 XP_005254845.1 3 449
XM_006720770.1 XP_006720833.1 3 449
XM_011522228.1 XP_011520530.1 3 449
XM_017022726.1 XP_016878215.1 3 553
GenBank mRNA AK294199.1 - 3 218
AK296234.1 - 3 558
AL832429.1 - 3 502
BC017708.1 - 3 504 3907760

View Full Product Details