Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033198.3 | NP_149975.1 | 9 | 1339 | |
GenBank mRNA | AB057723.1 | - | 9 | 966 | |
AF161427.1 | - | 3 | 200 | ||
AK025825.1 | - | 2 | 104 | ||
AK075428.1 | - | 9 | 994 | ||
AK127162.1 | - | 1 | 347 | ||
AK297046.1 | - | 8 | 858 | ||
AY359112.1 | - | 8 | 1234 | ||
BC001319.2 | - | 1 | 32 | ||
BC007301.1 | - | 7 | 723 | ||
BC069228.1 | - | 9 | 989 | 3944034 |