Gene Transcripts

Gene Symbol
WTAP
Entrez Gene ID
9589
Gene Name
Wilms tumor 1 associated protein
Gene Aliases
Mum2
Chromosome Location
Chr. 6: 159726680 - 159756319 on Build GRCh38
UniGene ID
Hs.446091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270531.1 NP_001257460.1 6 526
NM_001270532.1 NP_001257461.1 6 658
NM_001270533.1 NP_001257462.1 6 526
NM_004906.4 NP_004897.2 6 658
NM_152857.2 NP_690596.1 6 526
NM_152858.2 NP_690597.1 6 658
XM_017011514.1 XP_016867003.1 6 496
XM_017011515.1 XP_016867004.1 4 758
XM_017011516.1 XP_016867005.1 6 496
GenBank mRNA AF374416.1 - 6 539
AJ276706.1 - 5 306
AK127822.1 - 6 537
BC000383.2 - 6 517
BC004432.1 - 6 517
BC069192.1 - 6 494 4653831
BG702882.1 - 6 530
BQ948913.1 - 6 464
BX387163.2 - 6 508
CN284477.1 - 6 488
D14661.1 - 6 438
DA561768.1 - 6 532

View Full Product Details